Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Sodium Thiosulfate 0.122N, 4l

Catalog No. 504400685
Encompass_Preferred

Sodium Thiosulfate 0.122N, 4l Plastic Jug

Catalog No. 50-440-0685 Supplier LaMarKa Chemical Company Supplier No. 0000525916753108
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Thiosulfate 0.122N, 4l Plastic Jug

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.