Learn More
Organomation One Dozen 4in. X 19ga. Stainless Steel Needles
SDP
Catalog No. NC9433071
Encompass_Preferred
Set of one dozen luer lock needles. These needles are 4 inch by 19 gauge, with a 304 stainless steel shaft and a chrome plated brass hub.
Catalog No. NC9433071 Supplier Organomation Supplier No. NA0603
Set of one dozen luer lock needles. These needles are 4 inch by 19 gauge, with a 304 stainless steel shaft and a chrome plated brass hub.