Learn More
MilliporeSigma NA12/90 TIGHTENING TOOL
Catalog No. NC3962846
Encompass
Tightening Tool for Locking Ring on NovAseptic Valve. Compatible Valve Body NA NA12Compatible Valve Body NU NU050
Catalog No. NC3962846 Supplier MilliporeSigma Supplier No. NA12/90
Tightening Tool for Locking Ring on NovAseptic Valve. Compatible Valve Body NA NA12Compatible Valve Body NU NU050