Learn More
QA Bio Inc Ludger NA2 Glycan / G2 - 20µg
Supplier:  QA Bio Inc CNNA220U
Encompass
NA2 Glycan Synonyms: NA2 N-linked oligosaccharide. G2, A2G2, A2G(4)2. • Description: Asialo- bi-antennary complex-type N-glycan (oligosaccharide). • Molecular Weight: 1642 • Purity: >90% pure as assessed by a combination of 1 H-NMR and HPLC. • Sources: NA2 glycan is found on many mammalian glycoproteins including IgG, gamma globulins, and many serum glycoproteins. This product is typically purified from the oligosaccharide pool released from bovine serum by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution.
Catalog No. 50-223-6599