Learn More
QA Bio Inc Ludger NA2 Glycan, Procainamide labeled
Supplier:  QA Bio Inc CPROCNA201
Encompass
Procainamide labeled asialo, galactosylated biantennary oligosaccharide • LudgerTag procainamide labeled glycans can be analyzed by UHPLC, HPLC, ESI-MS, and LC-ESI-MS methods. Because of its improved ionization efficiency compared to 2-AB labeling it can permit identification of minor glycans (<1% relative peak area) by ESI-MS. • Description: Asialo- bi-antennary complex-type N-glycan (oligosaccharide). • Sources: NA2 glycan is found on many mammalian glycoproteins including IgG, gamma globulins, and many serum glycoproteins. This product is typically purified from the oligosaccharide pool released from bovine serum by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. • Molecular Weight: 1861 • Mass free dye: 235.33 • Mass Added to Glycan: 219.172 • Fluorescence: λex = 310 nm, λem = 370 nm. • Amount: 20 pmols • Purity: >90% pure as assessed by a combination of 1 H-NMR and HPLC.
Catalog No. 50-223-6619