Learn More
Agilent Technologies NA2G1F N-Glycan | 10 ug
Catalog No. NC9603959
Encompass
NA2G1F N-Glycan | Mol Wt: 1625.49 | 10 ug
Catalog No. NC9603959 Supplier Agilent Technologies Supplier No. GKC014301
NA2G1F N-Glycan | Mol Wt: 1625.49 | 10 ug