Learn More
MilliporeSigma NA51/90 TIGHTENING TOOL FOR LO
Catalog No. NC3962864
Encompass
Tightening Tool for Locking Ring on NovAseptic ValveCompatible Valve Body NA NA51Compatible Valve Body NU NU200
Catalog No. NC3962864 Supplier MilliporeSigma Supplier No. NA51/90
Tightening Tool for Locking Ring on NovAseptic ValveCompatible Valve Body NA NA51Compatible Valve Body NU NU200