Learn More
TARGETMOL CHEMICALS INC NAAD sodium salt 5MG
SDP
Catalog No. 502878063
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. NAAD sodium salt (NAAD Na salt) is a substrate of nicotinamide adenine dinucleotide synthase and can be used in studies about the specificity and kinetics of the substrate. Purity 98%
Catalog No. 50-287-8063 Supplier TARGETMOL CHEMICALS INC Supplier No. T335655MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. NAAD sodium salt (NAAD Na salt) is a substrate of nicotinamide adenine dinucleotide synthase and can be used in studies about the specificity and kinetics of the substrate. Purity 98%