Learn More
Cayman Chemical ε-NAADP+ (sodium salt), Purity: ≥95%, 3.8 mg
SDP
Catalog No. 502916929
Encompass_Preferred
A fluorescent derivative of NAADP; induces calcium release in sea urchin egg homogenates (EC50 = 5 µM); ex/em maxima at 275/410 nm, respectively, and its relative emission increases three-fold in DMSO
Catalog No. 50-291-6929 Supplier Cayman Chemical Supplier No. 384903.8 MG
A fluorescent derivative of NAADP; induces calcium release in sea urchin egg homogenates (EC50 = 5 µM); ex/em maxima at 275/410 nm, respectively, and its relative emission increases three-fold in DMSO