Learn More
Medchemexpress LLC Nab-Paclitaxel 50mg | 50mg
SDP
Supplier:  Medchemexpress LLC HYP9997450MG
Encompass_Preferred
Nab-Paclitaxel (Nanoparticle albumin-bound Paclitaxel) is an albumin-bound nanoparticle formulation of Paclitaxel (HY-B0015) Nab-Paclitaxel is composed of albumin and the active pharmaceutical ingredient Paclitaxel in which human albumin is used as an excipient to disperse and stabilize particles and carry the main drug Nab-Paclitaxel is a solvent-free taxane with higher response rates and improved tolerability Nab-Paclitaxel displays less toxicity and greater antitumor activity Nab-Paclitaxel is more readily available for tumor cell uptake in three rhabdomyosarcoma seven neuroblastoma cell lines and one ostersarcoma cell line Nab-Paclitaxel can be studied in cancer research for example breast cancer and solid tumors (The product specifications below only indicate the effective content of Paditaxel the actual albumin quality depends on the batch the ratio of each ingredient in this product is Paditaxel albumin -1 7 1 11)[1][2]
Catalog No. 50-003-22482