Learn More
Selleck Chemical LLC Nab-Paclitaxel-E1068-10MG
SDP
Catalog No. 503134428
Encompass_Preferred
Nab-Paclitaxel is a novel solvent-free paclitaxel in which the binding ratio of paclitaxel to human albumin is approximately 1 9 It is an anti-microtubule drug that promotes microtubule aggregation in tubulin dimers and inhibits microtubule depolymerization to stabilize the microtubule system Solutions are unstable and should be fresh-prepared
Catalog No. 50-313-4428 Supplier Selleck Chemical LLC Supplier No. E106810MG
Nab-Paclitaxel is a novel solvent-free paclitaxel in which the binding ratio of paclitaxel to human albumin is approximately 1 9 It is an anti-microtubule drug that promotes microtubule aggregation in tubulin dimers and inhibits microtubule depolymerization to stabilize the microtubule system Solutions are unstable and should be fresh-prepared