Learn More
Selleck Chemical LLC Nab-Paclitaxel-E1068-50MG
SDP
Catalog No. 503134429
Encompass_Preferred
Nab-Paclitaxel is a novel solvent-free paclitaxel in which the binding ratio of paclitaxel to human albumin is approximately 1 9 It is an anti-microtubule drug that promotes microtubule aggregation in tubulin dimers and inhibits microtubule depolymerization to stabilize the microtubule system Solutions are unstable and should be fresh-prepared
Catalog No. 50-313-4429 Supplier Selleck Chemical LLC Supplier No. E106850MG
Nab-Paclitaxel is a novel solvent-free paclitaxel in which the binding ratio of paclitaxel to human albumin is approximately 1 9 It is an anti-microtubule drug that promotes microtubule aggregation in tubulin dimers and inhibits microtubule depolymerization to stabilize the microtubule system Solutions are unstable and should be fresh-prepared