Learn More
Aviva Systems Biology Corp Nab2 Antibody - middle region (ARP36934_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36934_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nab2. It was validated on Western Blot. Nab2,Ngfi-A binding protein 2,AI451907; Nab2,NGFI-A-binding protein 2,NGFI-A-binding protein 2,EGR-1-binding protein 2,NGFI-A binding protein 2 (EGR-1 binding protein 2)The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI451907ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-548