Learn More
Aviva Systems Biology Corp Nab2 Antibody - middle region (ARP36933_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36933_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nab2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Nab2. Alternate names: AI451907ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-547