Learn More
Cayman Chemical Nα-Benzoyl-L-Arginine-pNA (hydrochloride), Purity: ≥98%, CAS: 21653-40-7, 100 mg
SDP
Catalog No. 502912176
Encompass_Preferred
A colorimetric substrate for trypsin and papain; enzymatic cleavage releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of trypsin or papain activity
Catalog No. 50-291-2176 Supplier Cayman Chemical Supplier No. 36331100 MG
A colorimetric substrate for trypsin and papain; enzymatic cleavage releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of trypsin or papain activity