Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nabht

Catalog No. 9123955G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-239-55G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-239-55G Supplier Sigma Aldrich Supplier No. 9123955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

NaBHT is an organic soluble base used for the Pd-catalyzed arylation of base-sensitve amines with aryl halides. NaBHT has also been show to be an efficient hydride source for the Pd-catalyzed reduction of aryl halides.Sodium Butylated Hydroxytoluene (NaBHT) as a New and Efficient Hydride Source for Pd-Catalysed Reduction Reactions

Specifications

CAS 191732-70-4
Molecular Formula C13H13N3O3
Linear Formula C13H13N3O3
MDL Number MFCD31692701
Quantity 100 mg
Synonym 3-(5-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione; E3 ligase ligand; Ligand for PROTAC(R) research
Molecular Weight (g/mol) 259.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.