Learn More
Medchemexpress LLC Nabumetone | 42924-53-8 | 99.9% | 1 ML
SDP
Catalog No. 5000408173
Encompass_Preferred
Nabumetone is an orally active non-acidic anti-inflammatory agent, acting as a potent and selective COX-2 inhibitor, and is the proagent of the active metabolite α-Demethylnaproxen. It can inhibit cancer cell proliferation and relieve gastric ulcers.
  • Orally active non-acidic anti-inflammatory agent
  • Potent and selective COX-2 inhibitor
  • Proagent of the active metabolite α-Demethylnaproxen
  • Inhibits cancer cell proliferation
  • Relieves gastric ulcers
Catalog No. 50-004-08173 Supplier Medchemexpress LLC Supplier No. HYB055910MM/1ML
Nabumetone is an orally active non-acidic anti-inflammatory agent, acting as a potent and selective COX-2 inhibitor, and is the proagent of the active metabolite α-Demethylnaproxen. It can inhibit cancer cell proliferation and relieve gastric ulcers.
  • Orally active non-acidic anti-inflammatory agent
  • Potent and selective COX-2 inhibitor
  • Proagent of the active metabolite α-Demethylnaproxen
  • Inhibits cancer cell proliferation
  • Relieves gastric ulcers