Learn More
Medchemexpress LLC Nabumetone-d3 (BRL14777-d3) | 1216770-08-9 | 98.8% | 1 MG
SDP
Catalog No. 5000408619
Encompass_Preferred
Nabumetone-d3 is the deuterium-labeled form of Nabumetone (HY-B0559), an orally active, non-acidic anti-inflammatory agent. It acts as a potent and selective COX-2 inhibitor and is a proagent for its active metabolite, α-Demethylnaproxen (HY-W086896). Nabumetone has been observed to inhibit cancer cell proliferation and reduce gastric ulcers. This compound is intended for research applications, such as a tracer or an internal standard in quantitative analyses like NMR, GC-MS, or LC-MS.
  • Deuterium-labeled form of nabumetone.
  • Orally active, non-acidic anti-inflammatory agent.
  • Potent and selective COX-2 inhibitor.
  • Proagent for its active metabolite.
  • Inhibits cancer cell proliferation.
  • Reduces gastric ulcers.
  • Intended for research applications.
  • Suitable as a tracer or internal standard in quantitative analyses like NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08619 Supplier Medchemexpress LLC Supplier No. HYB0559S1MG
Nabumetone-d3 is the deuterium-labeled form of Nabumetone (HY-B0559), an orally active, non-acidic anti-inflammatory agent. It acts as a potent and selective COX-2 inhibitor and is a proagent for its active metabolite, α-Demethylnaproxen (HY-W086896). Nabumetone has been observed to inhibit cancer cell proliferation and reduce gastric ulcers. This compound is intended for research applications, such as a tracer or an internal standard in quantitative analyses like NMR, GC-MS, or LC-MS.
  • Deuterium-labeled form of nabumetone.
  • Orally active, non-acidic anti-inflammatory agent.
  • Potent and selective COX-2 inhibitor.
  • Proagent for its active metabolite.
  • Inhibits cancer cell proliferation.
  • Reduces gastric ulcers.
  • Intended for research applications.
  • Suitable as a tracer or internal standard in quantitative analyses like NMR, GC-MS, or LC-MS.