Learn More
Aviva Systems Biology Corp NACA-RS ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP37646_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Naca-rs. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human Naca-rs. Alternate names: Gm1878ApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-676