Learn More
Cayman Chemical nacardIc AcId dIen 10mg
SDP
Supplier:  Cayman Chemical 2266310 MG
Encompass_Preferred
A polyunsaturated form of anacardic acid; has antibacterial activity against MRSA and S. mutans (MICs = 12.5 and 6.25 μg/ml, respectively); has schistosomicidal activity against adult S. mansoni worms at 100 μM; inhibits soybean lipoxygenase-1 in a time-dependent manner
Catalog No. 50-299-5577