Learn More
Cayman Chemical Anacardic Acid monoene, Purity: ≥95%, CAS: 22910-60-7, 10 mg
SDP
Catalog No. 502965811
Encompass_Preferred
A 2-hydroxy-6-alkylbenzoic acid in which the alkyl chain contains 15 carbons and is unsaturated at the 8 position; inhibits SUMOylation in vitro (IC50 = 3 µM) and in cells without affecting protein ubiquitination; directly binds the SUMO-activating enzyme E1, blocking the formation of the E1-SUMO intermediate
Catalog No. 50-296-5811 Supplier Cayman Chemical Supplier No. 1842210 MG
A 2-hydroxy-6-alkylbenzoic acid in which the alkyl chain contains 15 carbons and is unsaturated at the 8 position; inhibits SUMOylation in vitro (IC50 = 3 µM) and in cells without affecting protein ubiquitination; directly binds the SUMO-activating enzyme E1, blocking the formation of the E1-SUMO intermediate