Learn More
Cayman Chemical nacardIc AcId trIen 10mg
SDP
Catalog No. 502933834
Encompass_Preferred
A polyunsaturated form of anacardic acid; inhibits soybean lipoxygenase-1 (IC50 = 60 µM); inhibits superoxide production by xanthine oxidase by 82% at 88 µM; active against S. mutans, S. aureus, and MRSA ATCC 33591 (MICs = 1.56, 6.25, and 6.25 UG/ml, respectively); inhibits NADH oxidase (IC50 = 1.3 UG/ml); toxic to B. glabrata (LC50 = 0.35 ppm)
Catalog No. 50-293-3834 Supplier Cayman Chemical Supplier No. 2661110 MG
A polyunsaturated form of anacardic acid; inhibits soybean lipoxygenase-1 (IC50 = 60 µM); inhibits superoxide production by xanthine oxidase by 82% at 88 µM; active against S. mutans, S. aureus, and MRSA ATCC 33591 (MICs = 1.56, 6.25, and 6.25 UG/ml, respectively); inhibits NADH oxidase (IC50 = 1.3 UG/ml); toxic to B. glabrata (LC50 = 0.35 ppm)