Learn More
Cayman Chemical nacetrapb 10mg
SDP
Catalog No. 502986523
Encompass_Preferred
A CEPT inhibitor (IC50s = 17 and 15 nM for CETP-dependent transfer of cholesterol esters and triglycerides, respectively); decreases plasma levels of LDL and triglycerides, reduces atherosclerotic lesion sizes, and increases HDL levels in the E3L.CETP mouse model of atherosclerosis; increases HDL levels by 2-fold and lowers LDL and fasting plasma triglyceride levels in rhesus macaques at 150 mg/kg
Catalog No. 50-298-6523 Supplier Cayman Chemical Supplier No. 2155810 MG
A CEPT inhibitor (IC50s = 17 and 15 nM for CETP-dependent transfer of cholesterol esters and triglycerides, respectively); decreases plasma levels of LDL and triglycerides, reduces atherosclerotic lesion sizes, and increases HDL levels in the E3L.CETP mouse model of atherosclerosis; increases HDL levels by 2-fold and lowers LDL and fasting plasma triglyceride levels in rhesus macaques at 150 mg/kg