Learn More
Cayman Chemical N-acetyl-2,3-dehydro-2-Deoxyneuraminic Acid methyl ester, Purity: ≥95%, CAS: 25875-99-4, 100 mg
SDP
Catalog No. 502912943
Encompass_Preferred
A bacterial neuraminidase inhibitor (Ki = 48 µM for the A. sialophilus enzyme)
Catalog No. 50-291-2943 Supplier Cayman Chemical Supplier No. 36529100 MG
A bacterial neuraminidase inhibitor (Ki = 48 µM for the A. sialophilus enzyme)