Learn More
Cayman Chemical N-acetyl-2,3-dehydro-2-Deoxyneuraminic Acid, Purity: ≥95%, CAS: 24967-27-9, 10 mg
SDP
Catalog No. 502984661
Encompass_Preferred
An inhibitor of human neuraminidases (IC50s = 143, 43, 61, and 74 μM, for NEU 1, 2, 3, and 4, respectively); reduces latency to first seizure and increases seizure duration in a rat model of potassium-induced seizures
Catalog No. 50-298-4661 Supplier Cayman Chemical Supplier No. 1993910 MG
An inhibitor of human neuraminidases (IC50s = 143, 43, 61, and 74 μM, for NEU 1, 2, 3, and 4, respectively); reduces latency to first seizure and increases seizure duration in a rat model of potassium-induced seizures