Learn More
Cayman Chemical N-acetyl-2,3-dehydro-2-Deoxyneuraminic Acid methyl ester, Purity: ≥95%, CAS: 25875-99-4, 50 mg
SDP
Catalog No. 502911603
Encompass_Preferred
A bacterial neuraminidase inhibitor (Ki = 48 µM for the A. sialophilus enzyme)
Catalog No. 50-291-1603 Supplier Cayman Chemical Supplier No. 3652950 MG
A bacterial neuraminidase inhibitor (Ki = 48 µM for the A. sialophilus enzyme)