Learn More
Cayman Chemical N-acetyl-3,4-Methylenedioxymethcathinone, Purity: ≥95%, CAS: 1227293-15-3, 50 mg
SDP
Catalog No. 502892885
Encompass_Preferred
An acetylated analog of methylone, the MDMA-type designer drug that has been detected in products marketed as bath salts, plant food, and tablets; intended for forensic and research purposes
Catalog No. 50-289-2885 Supplier Cayman Chemical Supplier No. 900149250 MG
An acetylated analog of methylone, the MDMA-type designer drug that has been detected in products marketed as bath salts, plant food, and tablets; intended for forensic and research purposes