Learn More
Cayman Chemical N-acetyl-L-Cysteine ethyl ester, Purity: ≥95%, CAS: 59587-09-6, 100 mg
SDP
Catalog No. 502992467
Encompass_Preferred
An esterified form of NAC; has enhanced cell permeability compared to NAC in isolated perfused rat liver; prevents tert-butyl hydroperoxide-induced formation of methemoglobin in isolated human red blood cells at 1 mM; increases glutathione levels in rat liver, kidney, heart, testis, and brain at 50 mg/kg twice per day; reduces acetaminophen-induced increases in plasma AST, ALT, and LDH levels in rats
Catalog No. 50-299-2467 Supplier Cayman Chemical Supplier No. 30299100 MG
An esterified form of NAC; has enhanced cell permeability compared to NAC in isolated perfused rat liver; prevents tert-butyl hydroperoxide-induced formation of methemoglobin in isolated human red blood cells at 1 mM; increases glutathione levels in rat liver, kidney, heart, testis, and brain at 50 mg/kg twice per day; reduces acetaminophen-induced increases in plasma AST, ALT, and LDH levels in rats