Learn More
Cayman Chemical N-acetyl-S-farnesyl-L-Cysteine, Purity: ≥98%, CAS: 135304-07-3, 10 mg
SDP
Catalog No. 502896636
Encompass_Preferred
AFC is a synthetic substrate for the isoprenylated protein methyltransferase (also known as S-adenosylmethionine-dependent methyltransferase).{2306,2304} Because it is able to serve as a substrate for the methyltransferase, it effectively functions as an inhibitor of methylation of endogenous isoprenylated proteins.
Catalog No. 50-289-6636 Supplier Cayman Chemical Supplier No. 6327010 MG
AFC is a synthetic substrate for the isoprenylated protein methyltransferase (also known as S-adenosylmethionine-dependent methyltransferase).{2306,2304} Because it is able to serve as a substrate for the methyltransferase, it effectively functions as an inhibitor of methylation of endogenous isoprenylated proteins.