Learn More
Enzo Life Sciences Physiological saline (NaCl 0.9%) (endotoxin-free) (sterile) (1.5ml)
SDP
Catalog No. 502009346
Encompass_Preferred
Sterile and endotoxin-free physiological saline for use with TLRGRADE™ or endotoxin-free grade reagents. Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal. Long Term Storage: +4°C.
Catalog No. 50-200-9346 Supplier Enzo Life Sciences Supplier No. ALX505009LD15
Sterile and endotoxin-free physiological saline for use with TLRGRADE™ or endotoxin-free grade reagents. Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal. Long Term Storage: +4°C.