Learn More
Hach Company Certified Conductivity Standard Solution, 25 μS/cm, NaCl 25, 250 mL
Catalog No. 501998104 Shop All Hach Company Products
Encompass
Certified Conductivity Standard Solution with Certificate of Conformity and Traceability* reporting exact conductivity value with related expanded uncertainty (k=2). Glass bottle packaging. Sodium Chloride (NaCl) 25 Conductivity: 25.0 µS/cm ±5% (k=2) at 25°C, 250 mL bottle.*Available for download from www.hach.com: Support/Service, Certificate of Analysis menu using product part number and lot number.
Catalog No. 50-199-8104 Supplier Hach Company Supplier No. S51M013
Certified Conductivity Standard Solution with Certificate of Conformity and Traceability* reporting exact conductivity value with related expanded uncertainty (k=2). Glass bottle packaging. Sodium Chloride (NaCl) 25 Conductivity: 25.0 µS/cm ±5% (k=2) at 25°C, 250 mL bottle.*Available for download from www.hach.com: Support/Service, Certificate of Analysis menu using product part number and lot number.