Learn More
Cole-Parmer Cole-Parmer Replacement ionic strength adjustors, 5 M NaCl
Catalog No. 50633676 Shop All Cole-Parmer Products
Encompass_Preferred
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.
Catalog No. 50-633-676 Supplier Cole-Parmer Supplier No. 2750353
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.