Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics NaCl Diluent 9% 6 x 22 mL

Catalog No. 50589421 Shop All Roche Diagnostics Products
Encompass

NaCl Diluent 9% 6 x 22 mL

Catalog No. 50-589-421 Supplier Roche Diagnostics Supplier No. 06609406001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NaCl Diluent 9% 6 x 22 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.