Learn More
Hach Company Hach Saturated Potassium Chloride (KCl) Solution, 30 mL (S21M010)
Catalog No. NC2105740 Shop All Hach Company Products
Encompass
Saturated potassium chloride (KCl) solution supplied in a 30 mL container for electrochemical applications. It is intended for filling and storage of reference electrodes and for use in electrochemical measurements requiring a stable chloride environment.
  • Saturated potassium chloride aqueous solution.
  • 30 mL container.
  • For filling and storage of reference electrodes.
  • Used in electrochemical and conductivity measurements.
  • Consult the safety data sheet for handling and storage instructions.
This item is not returnable. View return policy
Catalog No. NC2105740 Supplier Hach Company Supplier No. S21M010
Saturated potassium chloride (KCl) solution supplied in a 30 mL container for electrochemical applications. It is intended for filling and storage of reference electrodes and for use in electrochemical measurements requiring a stable chloride environment.
  • Saturated potassium chloride aqueous solution.
  • 30 mL container.
  • For filling and storage of reference electrodes.
  • Used in electrochemical and conductivity measurements.
  • Consult the safety data sheet for handling and storage instructions.