Learn More
International Crystal NaCl Unpolished Disc- 25mmx5mm
Catalog No. NC9878481
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
NACL UNPOLISHED DISC 25MMX5MM. Unpolished crystal optics are also known as blanks. These windows are finely ground with flat parallel surfaces. As sold they are opaque. They can be water polished quickly to achieve 90% transmission. The transmission of water polished KBr, KCI and NaCl will be the same as Real Crystal IR Cards.These blanks can also be polished to precision optics. Blanks are ideal for mulls, smears and qualitative testing.
Catalog No. NC9878481 Supplier International Crystal Supplier No. 0002L131
This item has a minimum qty of 5 per supplier requirements.
NACL UNPOLISHED DISC 25MMX5MM. Unpolished crystal optics are also known as blanks. These windows are finely ground with flat parallel surfaces. As sold they are opaque. They can be water polished quickly to achieve 90% transmission. The transmission of water polished KBr, KCI and NaCl will be the same as Real Crystal IR Cards.These blanks can also be polished to precision optics. Blanks are ideal for mulls, smears and qualitative testing.