Learn More
Medchemexpress LLC HY-111505 10mg Medchemexpress, NAcM-OPT CAS:2089293-61-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11150510MG
Encompass_Preferred
Medchemexpress, HY-111505 10mg NAcM-OPT CAS:2089293-61-6 NAcM-OPT is an orally bioavailable cullin neddylation 1 (DCN1) inhibitor, which potently inhibits the DCN1-UBE2M interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8894