Learn More
Cayman Chemical N-acetyl-5-Aminosalicylic Acid, Purity: ≥98%, CAS: 51-59-2, 50 mg
SDP
Catalog No. 502937859
Encompass_Preferred
A metabolite of 5-ASA and sulfasalazine; reduces IFN-γ binding to colonic epithelial cells by 24% at 10 mM; scavenges DPPH radicals in a cell-free assay; inhibits hydroxy radical-stimulated DNA base hydroxylation; urinary levels have been used as a marker of 5-ASA adherence in patients with inflammatory bowel disease
Catalog No. 50-293-7859 Supplier Cayman Chemical Supplier No. 2761850 MG
A metabolite of 5-ASA and sulfasalazine; reduces IFN-γ binding to colonic epithelial cells by 24% at 10 mM; scavenges DPPH radicals in a cell-free assay; inhibits hydroxy radical-stimulated DNA base hydroxylation; urinary levels have been used as a marker of 5-ASA adherence in patients with inflammatory bowel disease