Learn More
Medchemexpress LLC HY-109008 5mg , Nacubactam CAS:1452458-86-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090085MG
Encompass_Preferred
HY-109008 5mg Nacubactam CAS: 1452458-86-4 Nacubactam (OP0595 free acid) is a potent non-β-lactam-β-lactamase inhibitor with activity against class A and class C β-lactamases. Nacubactam (OP0595 free acid) acts as a penicillin binding protein (PBP) 2-active antibacterial, and gives β-lactamase-independent potentiation of β-lactams targeting other PBPs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1276