Learn More
Medchemexpress LLC Nad+-13c5 ammonium | 53-84-9 | 98.9% | 1 MG
SDP
Catalog No. 5000408247
Encompass_Preferred
NAD+-13C5 (ammonium) is a 13C labeled NAD+ compound intended for research use only. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated into drug molecules primarily as tracers for quantitation during drug development. Shipping is at room temperature in the continental US, with variations elsewhere. Solid storage requires -20°C, sealed, away from moisture and light. If in solvent, store at -80°C for 6 months or -20°C for 1 month, also sealed and away from moisture and light.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08247 Supplier Medchemexpress LLC Supplier No. HYB0445S1MG
NAD+-13C5 (ammonium) is a 13C labeled NAD+ compound intended for research use only. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated into drug molecules primarily as tracers for quantitation during drug development. Shipping is at room temperature in the continental US, with variations elsewhere. Solid storage requires -20°C, sealed, away from moisture and light. If in solvent, store at -80°C for 6 months or -20°C for 1 month, also sealed and away from moisture and light.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS