Learn More
Research Products International Corp NAD 25 G
SDP
Catalog No. 502230352
Encompass_Preferred
β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.
Catalog No. 50-223-0352 Supplier Research Products International Corp Supplier No. N3011025.0
β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.