Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAD 299 hydrochloride, Tocris Bioscience™
Description
Selective, high affinity 5-HT1A receptor antagonist (Ki = 0.6 nM in vitro). Enhances the action of selective 5-HT reuptake inhibitors and reverses citalopram -induced (Cat. No. 1427) inhibition of serotonergic cell firing.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 354.85 |
| Percent Purity | >98% |
| Chemical Name or Material | NAD 299 hydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?