Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAD 299 hydrochloride, Tocris Bioscience™

Catalog No. p-4784175
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-8210 10 mg
32-825-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-8210 Supplier R&D Systems Supplier No. 3282/10
This item is not returnable. View return policy

Selective, high affinity 5-HT1A antagonist

Selective, high affinity 5-HT1A receptor antagonist (Ki = 0.6 nM in vitro). Enhances the action of selective 5-HT reuptake inhibitors and reverses citalopram -induced (Cat. No. 1427) inhibition of serotonergic cell firing.

Chemical Identifiers

CAS 184674-99-5
Molecular Formula C18H24ClFN2O2
Molecular Weight (g/mol) 354.85
InChI Key GSZJANKLCPHEEX-BTQNPOSSSA-N
Synonym nad 299 hydrochloride, 3r-3-dicyclobutylamino-8-fluoro-3,4-dihydro-2h-1-benzopyran-5-carboxamide hydrochloride
PubChem CID 90488823
IUPAC Name (3R)-3-[di(cyclobutyl)amino]-8-fluoro-3,4-dihydro-2H-chromene-5-carboxamide;hydrochloride
SMILES C1CC(C1)N(C2CCC2)C3CC4=C(C=CC(=C4OC3)F)C(=O)N.Cl

Specifications

Quantity 10 mg
Formula Weight 354.85
Percent Purity >98%
Chemical Name or Material NAD 299 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.