Learn More
Selleck Chemical LLC NAD S2518-1g
SDP
Catalog No. 503112976
Encompass_Preferred
NAD ( -DPN -NAD -Nicotinamide Adenine Dinucleotide nicotinamide adenine dinucleotide) is a coenzyme composed of ribosylnicotinamide 5 -diphosphate coupled to adenosine 5 -phosphate by pyrophosphate linkage
Catalog No. 50-311-2976 Supplier Selleck Chemical LLC Supplier No. S25181G
NAD ( -DPN -NAD -Nicotinamide Adenine Dinucleotide nicotinamide adenine dinucleotide) is a coenzyme composed of ribosylnicotinamide 5 -diphosphate coupled to adenosine 5 -phosphate by pyrophosphate linkage