Learn More
Medchemexpress LLC NAD+ | 53-84-9 | 100.0% | 50 G

Supplier: Medchemexpress LLC HYB044550G
NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage. It is an endogenous metabolite found widely in nature and is involved in numerous enzymatic reactions as an electron carrier, alternately oxidizing (NAD+) and reducing (NADH).
- High purity of 99.96%.
- Appears as a white to off-white solid.
- Molecular weight is 663.43.
- Attenuates glioma cell death.
- Suppresses declines of cysteine.
- Prevents RSL3-induced upregulation of ATF3.
- Improves expression of CD31 and VEGF in MI mice.
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
- Recommended storage in solvent: -80°C for 1 year, -20°C for 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.