Learn More
Research Products International Corp NAD [B-NAD] [B-Nicotinamide Adenine Dinucleotide, Oxidized free acid hydrate], 5 Grams
SDP
Catalog No. 50213554
Encompass_Preferred
β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.
Catalog No. 50-213-554 Supplier Research Products International Corp Supplier No. N301105.0
β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.