Learn More
Medchemexpress LLC NAD+ (beta-Nicotinamide Adenine Dinucleotide) | 53-84-9 | 99.96% | 1 ML
SDP
Catalog No. 5000408243
Encompass_Preferred
NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage. Found widely in nature, it is involved in numerous enzymatic reactions, serving as an electron carrier by being alternately oxidized (NAD+) and reduced (Nadide).
  • Involved in numerous enzymatic reactions as an electron carrier
  • White to off-white solid appearance
  • Available in solid and solution forms
Catalog No. 50-004-08243 Supplier Medchemexpress LLC Supplier No. HYB044510MM/1ML
NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage. Found widely in nature, it is involved in numerous enzymatic reactions, serving as an electron carrier by being alternately oxidized (NAD+) and reduced (Nadide).
  • Involved in numerous enzymatic reactions as an electron carrier
  • White to off-white solid appearance
  • Available in solid and solution forms