Learn More
Sigma Aldrich Fine Chemicals Biosciences NAD Grade I free acid
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10127973001
Encompass
Nicotinamide adenine dinucleotide (NAD) is a ubiquitously found electron carrier and a cofactor. NAD contains an adenylic acid and a nicotinamide-5AE-ribonucleotide group linked together by a pyrophosphate moiety. In NAD complexes the enzyme-cofactor interactions are highly conserved.
Catalog No. 50-290-2526