Learn More
Cell Biolabs Inc NAD+/NADH Assay Kit (Colorimetric), 100 assays
Supplier:  Cell Biolabs Inc MET5014
Encompass_Preferred
Cell Biolabs’ NAD+/NADH Assay Kit (Colorimetric) is a simple assay that measures NAD+ and NADH present in biological samples such as cell lysates or tissue extracts in a 96-well microtiter plate format. The kit is specific for NAD+, NADH, and their ratio. The kit will not detect NADP+ or NADPH. The total NAD+/NADH concentrations of unknown samples are determined by comparison with a known NAD+ standard. Determination of both NAD+ and NADH requires two separate samples for quantification. NAD+ and NADH do not need to be purified from samples, but rather can be extracted individually with a simple acid or base treatment prior to performing the assay. The kit has a detection sensitivity limit of approximately 4 nM NAD+. Each kit provides sufficient reagents to perform up to 100 assays, including blanks, NAD+ standards and unknown samples.
Catalog No. 50-125-2343