Learn More
Dojindo Molecular Technologies Inc NAD/NADH Assay Kit-WST (100TESTS), enables quantification of the amount of total NAD+/NADH, NAD+ and NADH in cells and measurement of their ratio.
SDP
Catalog No. 501903725
Encompass_Preferred
Recently, it has become clear that Sirtuin is linked to longevity and plays a role in NAD+ level regulation. Also Sirtuin has been recognized as a marker necessary to understanding biological states, such as obesity & diabetes, as well as cellular differentiation. Cell lysate from a cell culture can be easily prepared via Deproteinzation using the extraction buffer and filtration tubes found within this kit. Intracellular NADH levels can be quantified by heat treatment of cell lysate. Additionally, intracellular NAD+ levels can be determined by subtracting NADH levels from independently measured total NAD+/NADH levels. NAD/NADH Assay Kit-WST enables quantification of the amount of total NAD+/NADH, NADH and NAD+ in cells and measurement of their ratio. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)
Catalog No. 50-190-3725 Supplier Dojindo Molecular Technologies Inc Supplier No. N50910
Recently, it has become clear that Sirtuin is linked to longevity and plays a role in NAD+ level regulation. Also Sirtuin has been recognized as a marker necessary to understanding biological states, such as obesity & diabetes, as well as cellular differentiation. Cell lysate from a cell culture can be easily prepared via Deproteinzation using the extraction buffer and filtration tubes found within this kit. Intracellular NADH levels can be quantified by heat treatment of cell lysate. Additionally, intracellular NAD+ levels can be determined by subtracting NADH levels from independently measured total NAD+/NADH levels. NAD/NADH Assay Kit-WST enables quantification of the amount of total NAD+/NADH, NADH and NAD+ in cells and measurement of their ratio. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)