Learn More
Aat Bioquest Amplite™ Colorimetric NAD/NADH Ratio Assay Kit
SDP
Catalog No. 501953571
Encompass_Preferred
Nicotinamide adenine dinucleotide (NAD+) and nicotinamide adenine dinucleotide phosphate (NADP+) are two important cofactors found in cells. NADH is the reduced form of NAD+, and NAD+ is the oxidized form of NADH.
Catalog No. 50-195-3571 Supplier Aat Bioquest Supplier No. 15273
Nicotinamide adenine dinucleotide (NAD+) and nicotinamide adenine dinucleotide phosphate (NADP+) are two important cofactors found in cells. NADH is the reduced form of NAD+, and NAD+ is the oxidized form of NADH.